missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Resiniferatoxin, 99+%

$265.24 - $265.24

Chemical Identifiers

CAS 57444-62-9
Molecular Formula C37H40O9
Molecular Weight (g/mol) 628.718
MDL Number MFCD00135927
InChI Key DSDNAKHZNJAGHN-BWVJFJMLSA-N
Synonym resiniferatoxin
PubChem CID 57090162
SMILES CC1CC2(C3C4C1(C5C=C(C(=O)C5(CC(=C4)COC(=O)CC6=CC(=C(C=C6)O)OC)O)C)OC(O3)(O2)CC7=CC=CC=C7)C(=C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62458MCR View Documents Alfa Aesar™
J62458MCR
1mg Each for $265.24
 
Description

Description

Applications
Resiniferatoxin, a potent capsaicin-like stimulator of peripheral nociceptors in the neonatal rat tail in vitro. Resiniferatoxin is used as an anti-inflammatory, anti-viral and anti-psoriatic uses. Diterpene ester related to phorbol but apparently not tumorigenic activates protein kinase C

Solubility
Soluble in DMSO to 100mM and to 50mM in ethanol

Notes
Resiniferatoxin (RTX) is susceptible to slow degradation if exposed to air. Even at -20°C, Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep in a dry place. Keep away from strong oxidizing agents. RTX must be stored in an inert atmosphere for long-term stability.
Specifications

Chemical Identifiers

57444-62-9
628.718
DSDNAKHZNJAGHN-BWVJFJMLSA-N
57090162
C37H40O9
MFCD00135927
resiniferatoxin
CC1CC2(C3C4C1(C5C=C(C(=O)C5(CC(=C4)COC(=O)CC6=CC(=C(C=C6)O)OC)O)C)OC(O3)(O2)CC7=CC=CC=C7)C(=C)C

Specifications

White
57444-62-9
MFCD00135927
5371150
Soluble in DMSO to 100mM and to 50mM in ethanol
CC1CC2(C3C4C1(C5C=C(C(=O)C5(CC(=C4)COC(=O)CC6=CC(=C(C=C6)O)OC)O)C)OC(O3)(O2)CC7=CC=CC=C7)C(=C)C
57090162
Solid
Resiniferatoxin
1mg
C37H40O9
UN2811
resiniferatoxin
DSDNAKHZNJAGHN-BWVJFJMLSA-N
628.718
628.71
≥99%
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : CY1633700

Recommended Storage : −30° to −10°C