Learn More
Eppendorf™ Research™ Plus Single-Channel Multi-Pack PROMO
Essential bundles! Start, Expand, or Replace your pipettes with deals on 3- or 4-pack sets covering 2.5 to 1000 μL. Ultralight, adjustable volume, fully autoclavable bodies and spring-loaded tip cones deliver fatigue-free precision at every volume.
$1658.35 - $2983.53
Specifications
| Autoclavable | Autoclavable |
|---|---|
| Number of Channels | 1 |
| Product Line | Research Plus |
| Type | Pipette Package |
| Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity | |||||
|
05403151PM1
|
Eppendorf™
2231001167 |
2.5, 10, 100, 1000 μL |
Pack of 4 Each for $2,138.64
|
|
|||||
|
3116000015P
|
Eppendorf™
2231001168 |
2.5, 20, 200, and 1000 μL |
Each for $2,138.64
|
|
|||||
|
14285904PM
|
Eppendorf™
2231001169 |
10, 20, 200, 1000 μL |
Pack for $2,983.53
|
|
|||||
|
13684250PM
|
Eppendorf™
2231001170 |
10, 100, 1000 μL |
Each for $1,658.35
|
|
|||||
|
13684251PM1
|
Eppendorf™
2231001171 |
20, 200, 1000 μL |
Each for $1,658.35
|
|
|||||
Description
4 Pack Deals:
Eppendorf Research™ Plus 4-Pack and Carousel Stand Bundle (Option 1) Includes:
- 4 x Research™ Plus pipettes (2.5/10/100/1000 μL) with carousel stand
Eppendorf Research™ Plus 4-Pack and Carousel Stand Bundle (Option 2) Includes:
- 4 x Research™ Plus pipettes (2.5/20/200/1000 μL) with carousel stand
Eppendorf Research™ Plus 4-Pack and Carousel Stand Bundle (Option 3) Includes:
- 4 x Research™ Plus pipettes (10/20/200/1000 μL) with carousel stand
3 Pack Deals:
Eppendorf Research™ Plus 3-Pack Bundle (Option 1) Includes:
- 3 x Research™ Plus pipettes (10/100/1000 μL)
Eppendorf Research™ Plus 3-Pack Bundle (Option 2) Includes:
- 3 x Research™ Plus pipettes (20/200/1000 μL)
Eppendorf Research™ Plus Features:
- Volume adjustment feature allows desired volume to be reached in few quick turns
- Control button and ejectors low operating force are positioned for ergonomics
- 4-digit, magnifying-shape volume display
- Calibration display and adjustment opening for specific liquids and volumes
- Uses renowned PhysioCare Concept™ thus reduces strain on hand and arm during pipetting, maintains balance, minimal user exertion
Specifications
| Autoclavable | |
| 1 | |
| Research Plus | |
| Pipette Package |