missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Resazurin sodium salt, Alfa Aesar™

$38.03 - $38.03

Chemical Identifiers

CAS 62758-13-8
Molecular Formula C12H6NNaO4
Formula Weight 251.18
MDL Number MFCD00005036
Beilstein 3794971
Merck Index 14,8141
Synonym 7-Hydroxy-3H-phenoxazin-3-one-10-oxide sodium salt
Chemical Name or Material Resazurin sodium salt
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AAB2118703 SDS Alfa Aesar
B2118703
1g Each for $38.03
Specifications

Description

Applications
As indicator pH 3.8 (orange) to 6.5 (dark violet)Resazurin sodium salt acts as a blue non-fluorescent dye used especially in oxidation-reduction indicator of resazurin test for bacteria. It is employed as a fluorogenic oxidation-reduction indicator in a variety of cells, yeast and eukaryotes by flow cytometry, fluorescence microscopy and high-throughput screening. Further, it is used to characterize the metabolic activity of mitochnodria. It is also used to assay L-Glutamate and to measure the aerobic biodegradation of organic matter exists in effluents. In addition to this, it acts as an indicator for cell viability in mammalian cell cultures.

Solubility
Soluble in water, dimethylsulfoxide and ammonium hydroxide.

Notes
Incompatible with strong oxidizing agents. Store in a cool place.
Description & Specifications

Chemical Identifiers

62758-13-8
251.18
3794971
7-Hydroxy-3H-phenoxazin-3-one-10-oxide sodium salt
C12H6NNaO4
MFCD00005036
14,8141
Resazurin sodium salt

Specifications

62758-13-8
251.18
3794971
7-Hydroxy-3H-phenoxazin-3-one-10-oxide sodium salt
Resazurin sodium salt
C12H6NNaO4
MFCD00005036
14,8141
1g

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

RTECSNumber : SP7700000

TSCA : TSCA