Please login to your online account to display your discounted pricing

RepSox, Tocris™

Selective inhibitor of TGF-βRI

$218.74 - $915.10

Chemical Identifiers

Chemical Name or Material RepSox
Assay Percent Range >99%
Molecular Formula C17H13N5
Formula Weight 287.32
Synonym 2-(3-(6-Methylpyridine-2-yl)-1H-pyrazol-4-yl)-1, 5-naphthyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

374210

 
r & d systems
RDS374210
10mg Each for $218.74

374250

 
r & d systems
RDS374250
50mg Each for $915.10
Description & Specifications

Specifications

Formula Weight 287.32
Quantity 10mg
Chemical Name or Material RepSox
Assay Percent Range >99%
Molecular Formula C17H13N5
CAS 446859-33-2
Synonym 2-(3-(6-Methylpyridine-2-yl)-1H-pyrazol-4-yl)-1, 5-naphthyridine
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C