Please login to your online account to display your discounted pricing

Labconco™ Replacement Supply ULPA Filter

$1,453.32 - $1,453.32

Specifications

Material Borosilicate Microfiber
Weight (English) 5 lb.
Efficiency 99.999%
Filter Type ULPA
Weight (Metric) 2.26kg
View More Specs
Products
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity    

10-269-280

 
labconco™
LCN3859500
3 ft. Purifier™ Logic™ biosafety cabinet and Purifier™ Cell Logic™ biosafety cabinet Each for $1,453.32
Description & Specifications

Specifications

Material Borosilicate Microfiber
Weight (English) 5 lb.
Efficiency 99.999%
Filter Type ULPA
Weight (Metric) 2.26kg