Please login to your online account to display your discounted pricing
Replacement Reagents for Oakton™ Colorimeters
Supply of reagent to perform 50 tests
Manufacturer: oakton instruments  WD3564566
 View more versions of this product
Catalog No. 13-300-258
  • / Pack
Description & Specifications
Specifications
No. of Tests 50.0
For Use With (Equipment) C201, C301, C401 Oakton™ colorimeters
Certifications/Compliance EPA, DPD
Product Type Total Chlorine Reagent
Range 2 to 6ppm