Please login to your online account to display your discounted pricing
Replacement Reagents for Oakton™ Colorimeters
Supply of reagent to perform 50 tests
$79.11 - $80.94
Specifications
Certifications/Compliance EPA, DPD
No. of Tests 50.0
View More Specs
Products
Catalog Number Mfr. No. For Use With (Equipment) Product Type Price Quantity    
13-300-257
oakton instruments
WD3564564
C201, C301, C401 Oakton™ colorimeters Free Chlorine Reagent Pack for $80.94
13-300-258
oakton instruments
WD3564566
C201, C301, C401 Oakton™ colorimeters Total Chlorine Reagent Pack for $79.11
Description & Specifications
Specifications
Certifications/Compliance EPA, DPD
No. of Tests 50.0
Uses EPA-approved DPD method for chlorine testing