Please login to your online account to display your discounted pricing
Replacement Parts Kits for Drummond Microdispensers
Manufacturer: drummond scientific co  3000010K**
 View more versions of this product
Catalog No. DRM3000010K
  • / Each
Description & Specifications
Specifications
Includes 2 Plungers, 2 O-rings, 1 collet, 1 coil spring, 1 spring guide
For Use With (Equipment) 2 to 10μL Drummond™ microdispenser
Product Type Microdispenser Replacement Parts Kit