Please login to your online account to display your discounted pricing
Thermo Scientific™ Replacement HEPA Filters and Cleaning Kits
Help ensure measurable Class 100 air quality performance with Thermo Scientific™ Replacement HEPA Filters and Cleaning Kits.
$129.40 - $453.83
Products
Catalog Number Mfr. No. Description For Use With Price Quantity    
15-497-025
thermo scientific™
190651
3110 Kit: HEPA Filter, gasket, misc parts 3110 Series Incubators Each for $305.64
15-467-190
thermo scientific™
FOR760175
Replacement HEPA Filter 3110 Series, 310 Series (w/ HEPA option), Steri-Cycle Series incubators Each for $129.40
15-497-023
thermo scientific™
760209
Main HEPA Filter, Value pack (4/pk) 8000 Series models Pack of 4 Each for $453.83
Description & Specifications
Help ensure measurable Class 100 air quality performance with Thermo Scientific™ Replacement HEPA Filters and Cleaning Kits.
HEPA Filters
  • 99.97% filtering at 0.3μm
  • Compatible with all Thermo Scientific™ Forma™ Series II 3110, Thermo Scientific™ Steri-Cycle™, and Forma™ 310 Series (with HEPA filtration system option) incubators
  • Main chamber filter available as a stand-alone or packaged in a group with the other chamber in-line filters
Decontamination Kits
  • Consists of filters for difficult-to-clean chambers and gaskets (except door) and tubing
  • Reduces downtime and labor spent on decontamination