missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Repaglinide 98.0+%, TCI America™

Manufacturer:  Tci America R01791G

 View more versions of this product

Catalog No. R01791G



Specifications

Specifications

Repaglinide
C27H36N2O4
(S)-(+)-2-Ethoxy-4-[N-[1-(2-piperidinophenyl)-3-methyl-1-butyl]aminocarbonylmethyl]benzoic Acid
CCOC1=C(C=CC(=C1)CC(=O)NC(CC(C)C)C2=CC=CC=C2N3CCCCC3)C(=O)O
452.595
CHEBI:8805
Crystal-Powder at 20°C
133°C
1g
135062-02-1
MFCD00906179
FAEKWTJYAYMJKF-QHCPKHFHSA-N
2-ethoxy-4-[2-[[(1S)-3-methyl-1-(2-piperidin-1-ylphenyl)butyl]amino]-2-oxoethyl]benzoic acid
65981
452.60
≥98.0% (HPLC,T)
White-Yellow

Chemical Identifiers

135062-02-1
452.595
FAEKWTJYAYMJKF-QHCPKHFHSA-N
65981
2-ethoxy-4-[2-[[(1S)-3-methyl-1-(2-piperidin-1-ylphenyl)butyl]amino]-2-oxoethyl]benzoic acid
C27H36N2O4
MFCD00906179
(S)-(+)-2-Ethoxy-4-[N-[1-(2-piperidinophenyl)-3-methyl-1-butyl]aminocarbonylmethyl]benzoic Acid
CHEBI:8805
CCOC1=C(C=CC(=C1)CC(=O)NC(CC(C)C)C2=CC=CC=C2N3CCCCC3)C(=O)O
Safety and Handling

Safety and Handling

RTECSNumber : DI0876305

TSCA : No

Recommended Storage : Refrigerator

Documents