Please login to your online account to display your discounted pricing

Alfa Aesar™ Renin Inhibitor Peptide


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66928MA

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66928MA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 10mg
Format Solid
Sequence His-Pro-Phe-His-Leu-D-Leu-Val-Tyr-NH2
Regulatory Status RUO
Storage Requirements -30° to -10°C
Product Type Renin Inhibitor Peptide
Molecular Weight 1024.23
Purification Method HPLC