missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Renin Inhibitor Peptide

For cellular and molecular biology applications

$256.95 - $459.16

Specifications

Product Type Renin Inhibitor Peptide
Molecular Weight 1024.23
Storage Requirements -30° to -10°C
Format Solid
Sequence His-Pro-Phe-His-Leu-D-Leu-Val-Tyr-NH2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66928MA View Documents Alfa Aesar™
J66928MA
10mg Each for $256.95
 
AAJ66928MB View Documents Alfa Aesar™
J66928MB
25mg Each for $459.16
 
Specifications

Specifications

Renin Inhibitor Peptide
-30° to -10°C
His-Pro-Phe-His-Leu-D-Leu-Val-Tyr-NH2
HPLC
1024.23
Solid
RUO

For Research Use Only