Please login to your online account to display your discounted pricing

Remoxipride hydrochloride, Tocris


Selective D2-like antagonist

Quantity 10mg

Manufacturer: r & d systems  091610

 View more versions of this product

Catalog No. 091610

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 407.73
Chemical Name or Material Remoxipride hydrochloride
CAS 73220-03-8
Molecular Formula C16H23BrN2O3·HCl
Assay Percent Range >98%
Synonym (S)-(-)-3-Bromo-N-[(1-ethyl-2-pyrrolidinylmethyl]2, 6-dimethoxybenzamide hydrochloride
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material Remoxipride hydrochloride
Assay Percent Range >98%
Molecular Formula C16H23BrN2O3·HCl
Formula Weight 407.73
Synonym (S)-(-)-3-Bromo-N-[(1-ethyl-2-pyrrolidinylmethyl]2, 6-dimethoxybenzamide hydrochloride
Accessories

Store at Room Temperature