Please login to your online account to display your discounted pricing

Remoxipride hydrochloride, Tocris

Selective D2-like antagonist

$181.19 - $762.58

Chemical Identifiers

Chemical Name or Material Remoxipride hydrochloride
Assay Percent Range >98%
Molecular Formula C16H23BrN2O3·HCl
Formula Weight 407.73
Synonym (S)-(-)-3-Bromo-N-[(1-ethyl-2-pyrrolidinylmethyl]2, 6-dimethoxybenzamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

091610

 
r & d systems
091610
10mg Each for $181.19

091650

 
r & d systems
091650
50mg Each for $762.58
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 407.73
Chemical Name or Material Remoxipride hydrochloride
Assay Percent Range >98%
Molecular Formula C16H23BrN2O3·HCl
CAS 73220-03-8
Synonym (S)-(-)-3-Bromo-N-[(1-ethyl-2-pyrrolidinylmethyl]2, 6-dimethoxybenzamide hydrochloride
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature