Thermo Scientific™ Remel™ UVM Modified Listeria Enrichment Broth
Thermo Scientific™ Remel UVM Modified Listeria Enrichment Broth is a selective enrichment for isolation of Listeria monocytogenes.
Manufacturer:  Thermo Scientific™ RE112850
 View more versions of this product
Catalog No. R112850
Description
Description
Thermo Scientific™ Remel UVM Modified Listeria Enrichment Broth is a selective enrichment for isolation of Listeria monocytogenes.
Ordering Information:
REF R117870 is polypropylene vial with screw cap.
Specifications
Specifications
Industrial Reference: COMPF.
BB - 225mL
10 x 225mL
UVM Modified Listeria Enrichment Broth
10/Pk.
Broth
Documents
Not intended for IVD use.