Thermo Scientific™ Remel™ Trichrome Stain Kit
Thermo Scientific™ Remel Trichrome Stain Kit is used for trichrome staining.
$1,218.90 - $1,218.90
Specifications
Type Kit
Includes 250mL Trichrome Stain (Wheatley); 3 x 250mL Ethanol 70%; 250mL Acid Ethanol 90%; 2 x 250mL Ethanol 95%; 2 x 250mL Xylene-S; 5 x 0.75mL Lugol's Iodine ampules.
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Type Price Quantity  
Catalog Number Mfr. No. Type Price Quantity  
R40217 Thermo Scientific™
RE40217
Kit Each for $1,218.90
Specifications
Description
Thermo Scientific™ Remel Trichrome Stain Kit is used for trichrome staining.
Includes:
250mL Trichrome Stain (Wheatley); 3 x 250mL Ethanol 70%; 250mL Acid Ethanol 90%; 2 x 250mL Ethanol 95%; 2 x 250mL Xylene-S; 5 x 0.75mL Lugol's Iodine ampules.
Description & Specifications
Specifications
Kit
250mL Trichrome Stain (Wheatley); 3 x 250mL Ethanol 70%; 250mL Acid Ethanol 90%; 2 x 250mL Ethanol 95%; 2 x 250mL Xylene-S; 5 x 0.75mL Lugol's Iodine ampules.