Thermo Scientific™ Remel™ Non-Calibrated Inoculating Loops
Thermo Scientific™ Remel Non-Calibrated Inoculating Loops are twisted nichrome without handle.
$54.90 - $256.40
Products ${productFamilyLength}
Catalog Number Mfr. No. Color Format Packaging Price Quantity  
Catalog Number Mfr. No. Color Format Packaging Price Quantity  
R503153 Thermo Scientific™
RE503153
Green 3mm 1 Each Each for $54.90
R503151 Thermo Scientific™
RE503151
White 2mm 1 Each Each for $54.90
R503515 Thermo Scientific™
RE503515
5mm 25 Per Pack Pack of 25 Each for $180.60
R503152 Thermo Scientific™
RE503152
White 2mm 5 Per Pack Pack of 5 Each for $180.60
R503512 Thermo Scientific™
RE503512
2mm 25 Per Pack Pack of 25 Each for $180.60
R503156 Thermo Scientific™
RE503156
Yellow 5mm 5 Per Pack Pack of 5 Each for $256.40
R503154 Thermo Scientific™
RE503154
Green 3mm 5 Per Pack Pack of 5 Each for $256.40
R503155 Thermo Scientific™
RE503155
Yellow 5mm 1 Each Each for $54.90
R503513 Thermo Scientific™
RE503513
3mm 25 Per Pack Pack of 25 Each for $256.40
Specifications
Description
Thermo Scientific™ Remel Non-Calibrated Inoculating Loops are twisted nichrome without handle.
  • Twisted nichrome made
  • With or without insulated handle
  • Available in different color codes