Thermo Scientific™ Remel™ MRS Broth (Man, Rogosa, Sharpe Broth)
For the cultivation of Lactobacillus spp. and differentiation of gram-positive cocci.
Manufacturer:  Thermo Scientific™ RE061426
 View more versions of this product
Catalog No. R061426
Description
Description
Thermo Scientific™ Remel MRS Broth (Man, Rogosa, Sharpe Broth) is for the cultivation of Lactobacillus spp. and differentiation of gram-positive cocci.
Specifications
Specifications
Industrial Reference: COMPF, IDF.
20 x 7mL
MRS Broth
15mm x 103mm Durham Tube
20/Pk.
Broth