Thermo Scientific™ Remel™ MRS Broth (Man, Rogosa, Sharpe Broth)
For the cultivation of Lactobacillus spp. and differentiation of gram-positive cocci.
$449.18 - $542.61
Specifications
Certifications/Compliance Industrial Reference: COMPF, IDF.
Description MRS Broth
Packaging 20/Pk.
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
R061426
View Documents
Thermo Scientific™
RE061426
20 x 7mL
Pack of 20 Each for $542.61
R061428
View Documents
Thermo Scientific™
RE061428
20 x 2
Pack of 20 Each for $449.18
Description
Description
Thermo Scientific™ Remel MRS Broth (Man, Rogosa, Sharpe Broth) is for the cultivation of Lactobacillus spp. and differentiation of gram-positive cocci.
Specifications
Specifications
Industrial Reference: COMPF, IDF.
MRS Broth
20/Pk.