Thermo Scientific™ Remel™ LB Agar w/Ampicillin (100μg/mL) (Luria-Bertani)
Use for the cultivation and maintenance of Escherichia coli for genetic and molecular studies.
$160.60 - $160.60
Specifications
Description | LB (Luria-Bertani) Agar w/Ampicillin 100micrograms/mL |
---|---|
Packaging | 10/Pk. |
Catalog Number | Mfr. No. | Format | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Format | Price | Quantity | |||||
R110204
|
Thermo Scientific™
RE110204 |
12mm x 85mm Monoplate |
Pack of 10 Each for $160.60
|
|
Description
Thermo Scientific™ Remel LB Agar with Ampicillin (100μg/mL) (Luria-Bertani) is used for the cultivation and maintenance of Escherichia coli for genetic and molecular studies.
Specifications
LB (Luria-Bertani) Agar w/Ampicillin 100micrograms/mL | |
10/Pk. |