missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Hippurate Broth (3mL)

Hippurate Broth is used to determine the ability of an organism to hydrolyze sodium hippurate.

Manufacturer:  Thermo Scientific™ RE061152

 View more versions of this product

Catalog No. R061152



Description

Description

Determine the ability of an organism to hydrolyze sodium hippurate by enzymatic action using Thermo Scientific™ Remel™ Hippurate Broth. Heart muscle infusion and animal tissue peptone supply the variety of nutrients required for the growth of the majority of bacterial species. Sodium chloride maintains osmotic equilibrium. In 1922, Ayers and Rupp used an enriched medium containing hippuric acid to investigate the ability of bovine and human β-hemolytic streptococci to split hippuric acid into benzoic acid and glycine1. In 1951, Leuthardt described hippuricase, the enzyme responsible for the hydrolysis of hippurate2.

Requires:

Hippurate Hydrolysis Reagent (12% Ferric Chloride), Part No. R21221

Specifications

Specifications

Hippurate Broth Sodium Hippurate
Broth
20/Pk.
15mm x 103mm Tube
Broth
SDS
Documents
Product Certifications
Promotions

Promotions