Learn More
Thermo Scientific™ Remel™ Hippurate Broth (Sodium Hippurate)
Thermo Scientific™ Remel Hippurate Broth (Sodium Hippurate) is used for determination of enzymatic hydrolysis of sodium hippurate.
$266.19 - $266.19
Specifications
Description | Hippurate Broth Sodium Hippurate |
---|---|
Format | 15mm x 103mm Tube |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
R061152
|
Thermo Scientific™
RE061152 |
20/Pk. |
Pack of 20 Each for $266.19
|
|
Description
Thermo Scientific™ Remel Hippurate Broth (Sodium Hippurate) is used for determination of enzymatic hydrolysis of sodium hippurate.
Requires:
Hippurate Hydrolysis Reagent (12% Ferric Chloride), Part No. R21221
Specifications
Hippurate Broth Sodium Hippurate | |
15mm x 103mm Tube |