Please login to your online account to display your discounted pricing
Thermo Scientific™ Remel™ Eugonic Broth Modified
Provides cultivation of a variety of microorganisms including Neisseria, Haemophilus and anaerobic bacteria.
Manufacturer: thermo scientific™  RE07105
 View more versions of this product
Catalog No. R07105
  • / Pack of 20 Each
Description & Specifications
Specifications
Type Broth
Format 16mm x 125mm Tube
Description Eugonic Broth Modified
Packaging 20/Pk.
Quantity 20 x 10mL
Thermo Scientific™ Remel Eugonic Broth Modified provides cultivation of a variety of microorganisms including Neisseria, Haemophilus and anaerobic bacteria.