Thermo Scientific™ Remel™ Enhanced Gram Stain Kit
Thermo Scientific™ enhanced gram stain reagents are balanced to give a distinct gram-positive reaction and are less prone to over decolorization.
$328.30 - $328.30
Specifications
Type 250mL/Bottle
Includes 1 Gram Crystal Violet, Enhanced; 1 Gram Iodine, Enhanced; 1 Gram Decolorizer/Fixative, Enhanced; 1 Gram Safranin, Enhanced; 1 Gram Enhancer.
Quantity Kit
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Quantity Type Price Quantity  
Catalog Number Mfr. No. Includes Quantity Type Price Quantity  
R40240 Thermo Scientific™
RE40240
1 Gram Crystal Violet, Enhanced; 1 Gram Iodine, Enhanced; 1 Gram Decolorizer/Fixative, Enhanced; 1 Gram Safranin, Enhanced; 1 Gram Enhancer. Kit 250mL/Bottle Pack for $328.30
Specifications
Description
Thermo Scientific™ Remel enhanced gram stain reagents are balanced to give a distinct gram-positive reaction and are less prone to over decolorization.
  • Gram Enhancer is used to reduce the red color of background debris to give better contrast to small gram-negative microorganisms
Includes:
1 Gram Crystal Violet, Enhanced; 1 Gram Iodine, Enhanced; 1 Gram Decolorizer/Fixative, Enhanced; 1 Gram Safranin, Enhanced; 1 Gram Enhancer.
Description & Specifications
Specifications
250mL/Bottle
1 Gram Crystal Violet, Enhanced; 1 Gram Iodine, Enhanced; 1 Gram Decolorizer/Fixative, Enhanced; 1 Gram Safranin, Enhanced; 1 Gram Enhancer.
Kit