Thermo Scientific™ Remel™ Enhanced Gram Decolorizer/Fixative
Thermo Scientific™ enhanced gram stain reagents are balanced to give a distinct gram-positive reaction and are less prone to over decolorization.
$308.58 - $626.20
Description
Thermo Scientific™ Remel enhanced gram stain reagents are balanced to give a distinct gram-positive reaction and are less prone to over decolorization.
Specifications
Decolorizer/Fixative |