Thermo Scientific™ Remel™ Contact Plate Sterile Sabouraud Dextrose Agar w/Lecithin, Polysorbate 80 (pH 5.6) (Irradiated)
Thermo Scientific™ Remel Contact Plate Sterile Sabouraud Dextrose Agar with Lecithin, Polysorbate 80 (pH 5.6, Irradiated) is used for cultivation of yeast and fungi.
$87.30 - $87.30
Specifications
Description Contact Plates; Sabouraud Dextrose Agar w/Lecithin; Gamma irradiated
Format Contact - Double Bag
View More Specs
Disclaimers:
Not intended for IVD use.
Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Price Quantity  
Catalog Number Mfr. No. Packaging Price Quantity  
R111805 Thermo Scientific™
RE111805
10/Pk. Pack of 10 Each for $87.30
Specifications
Description
Thermo Scientific™ Remel Contact Plate Sterile Sabouraud Dextrose Agar with Lecithin, Polysorbate 80 (pH 5.6) (Irradiated) is used for the cultivation of yeast and fungi.
  • Contact Plates Sterile are double bagged plus a 3rd bag is enclosed for customer use (Double + Bag)
  • These products are manufactured in a clean room and validated sterile by gamma radiation
  • The Contact Plates Sterile listed are made using GripFit™ technology, which ensures the lid remains in place during handling and yet can be easily removed during environmental monitoring
Description & Specifications
Specifications
Contact Plates; Sabouraud Dextrose Agar w/Lecithin; Gamma irradiated
Contact - Double Bag