Thermo Scientific™ Remel™ Contact II Tryptic Soy Agar w/Lecithin, Polysorbate 80
Thermo Scientific™ Remel Contact II Tryptic Soy Agar w/Lecithin, Polysorbate 80 is used for the cultivation of bacteria, yeast, and fungi.
$151.13 - $151.13
Specifications
Description | Contact II Tryptic Soy Agar w/Lecithin, Polysorbate 80 |
---|---|
Format | Contact - Double Bag |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
R111201
|
Thermo Scientific™
RE111201 |
20/Pk. |
Pack of 20 Each for $151.13
|
|
Description
Thermo Scientific™ Remel Contact II Tryptic Soy Agar w/Lecithin, Polysorbate 80 is used for the cultivation of bacteria, yeast and fungi.
- Manufactured in a clean room environment to provide the highest level of sterility assurance
- Made using GripFit™ technology, which ensures the lid remains in place during handling and yet can be easily removed during environmental monitoring
- Contact I (Single Bag) and Contact II (Double Bag) are manufactured in a clean room environment
- This provides the highest level of sterility assurance with no compromise of growth characteristics
- Each lot is tested to ensure consistent, reproducible results
Specifications
Contact II Tryptic Soy Agar w/Lecithin, Polysorbate 80 | |
Contact - Double Bag |
Not intended for IVD use.