Thermo Scientific™ Reusable Calibrated Inoculating Loops
Select, inoculate or transfer a well-defined microbiological sample volume on media plates with these reusable calibrated inoculating loops.
Supplier: Thermo Scientific™ RE503630
Description
Select, inoculate or transfer microbiological sample on media with Thermo Scientific™ Remel™ Reusable Calibrated Inoculating Loops. The gentle, malleable loops offer precise control when selecting or streaking inoculum, without cutting or tearing agar surfaces. The loops are nontoxic, lightweight and are color-coded for easy-recognition and streamlined specimen handling.
- Platinum, twisted nichrome or plastic
- With or without insulated handle
- Some are color coded
Specifications
Plastic, Platinum, Nichrome | |
0.01 μL | |
Inoculating Loop Twisted Nichrome | |
Each |