Thermo Scientific™ Remel™ Butterfield's Phosphate Buffer
Thermo Scientific™ Remel Butterfield's Phosphate Buffer (pH 7.2) is used as a diluent in microbial limit testing of pharmaceutical products and food testing.
Manufacturer:  Thermo Scientific™ RE112090
 View more versions of this product
Catalog No. R112090
Description
Description
Thermo Scientific™ Remel Butterfield's Phosphate Buffer (pH 7.2) is used as a diluent in microbial limit testing of pharmaceutical products and food testing.
Not intended for IVD use.
Specifications
Specifications
Industrial Reference: BAM, USDA, USP.
20 x 400mL
Phosphate Buffer, Butterfield's
Clear Square Bottle - 400mL
20/Pk.
Liquid
Documents