Thermo Scientific™ Remel™ BactiDrop Lactophenol Aniline Blue
Remel BactiDrop Lactophenol Aniline Blue is for staining fungal elements.
$217.40 - $217.40
Specifications
Type BactiDrop Lactophenol Aniline Blue
Quantity 50 Pk.
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Type Price Quantity  
Catalog Number Mfr. No. Quantity Type Price Quantity  
R21526 Thermo Scientific™
RE21526
50 Pk. BactiDrop Lactophenol Aniline Blue Pack of 50 Each for $217.40
Specifications
Description
Remel BactiDrop Lactophenol Aniline Blue is for staining fungal elements.
Ordering Alerts:
BactiDrop reagents are supplied in crushable ampules encased in a protective plastic sheath with a convenient dropper dispenser tip. BactiDrop stains are in crushable ampules enclosed in a protective plastic sheath with a convenient dropper dispenser tip.
Description & Specifications
Specifications
BactiDrop Lactophenol Aniline Blue
50 Pk.