Thermo Scientific™ Remel™ BactiDrop Calcofluor White
A fluorescent stain for fungi, Microsporidium, Acanthamoeba, and Pneumocystis.
$217.40 - $217.40
Specifications
Type BactiDrop Calcofluor White
Quantity 50 Pk.
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Type Price Quantity  
Catalog Number Mfr. No. Type Price Quantity  
R21507 Thermo Scientific™
RE21507
BactiDrop Calcofluor White Pack of 50 Each for $217.40
Specifications
Description
Remel BactiDrop Calcofluor White is a fluorescent stain for fungi, Microsporidium, Acanthamoeba, and Pneumocystis.
Ordering Alerts:
BactiDrop reagents are supplied in crushable ampules encased in a protective plastic sheath with a convenient dropper dispenser tip. BactiDrop stains are in crushable ampules enclosed in a protective plastic sheath with a convenient dropper dispenser tip.
Description & Specifications
Specifications
BactiDrop Calcofluor White
50 Pk.