Thermo Scientific™ Remel™ BactiDrop Acridine Orange
Thermo Scientific™ Remel BactiDrop Acridine Orange is a fluorescent stain for detecting bacteria in body fluids.
$217.40 - $217.40
Specifications
Type BactiDrop Acridine Orange
Quantity 50 Pk.
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Type Price Quantity  
Catalog Number Mfr. No. Quantity Type Price Quantity  
R21502 Thermo Scientific™
RE21502
50 Pk. BactiDrop Acridine Orange Pack of 50 Each for $217.40
Specifications
Description
Remel BactiDrop Acridine Orange is a fluorescent stain for detecting bacteria in body fluids.
Ordering Alerts:
BactiDrop reagents are supplied in crushable ampules encased in a protective plastic sheath with a convenient dropper dispenser tip. BactiDrop stains are in crushable ampules enclosed in a protective plastic sheath with a convenient dropper dispenser tip.
Description & Specifications
Specifications
BactiDrop Acridine Orange
50 Pk.