Please login to your online account to display your discounted pricing

Remacemide hydrochloride, Tocris

NMDA antagonist; blocks ion channel and allosteric modulatory site

$218.74 - $915.10

Chemical Identifiers

Chemical Name or Material Remacemide hydrochloride
Assay Percent Range >99%
Molecular Formula C17H20N2O·HCl
Formula Weight 304.82
Synonym 2-Amino-N-(1-methyl-1, 2-diphenylethyl)acetamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

162250

 
r & d systems
RDS162250
50mg Each for $915.10

162210

 
r & d systems
RDS162210
10mg Each for $218.74
Description & Specifications

Specifications

Formula Weight 304.82
Quantity 50mg
Chemical Name or Material Remacemide hydrochloride
Assay Percent Range >99%
Molecular Formula C17H20N2O·HCl
CAS 111686-79-4
Synonym 2-Amino-N-(1-methyl-1, 2-diphenylethyl)acetamide hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature