Stemgent™ Alvetex™ Scaffold Multiwell Plate Formats
Alvetex forms highly porous polystyrene scaffold for 3D cell culture, providing 3D space in which cells can invade, proliferate and grow
$14934.03 - $52430.71
Catalog Number | Mfr. No. | No. of Wells | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | No. of Wells | Quantity | Price | Quantity | ||||
AVP00280
|
Stemgent™
AVP00280 |
12 | 80 |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AVP00680
|
Stemgent™
AVP00680 |
24 | 80 |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AVP009320
|
Stemgent™
AVP009320 |
96 | 320 |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Sterilized using gamma radiation
- Individually sterile blister-packed plates
- Vertical axis of 3D enables additional layers of cells to form complex 3D interactions with adjacent cells simulating structure of tissue