Stemgent™ Alvetex™ Scaffold Multiwell Plate Formats DFS Item
Alvetex forms highly porous polystyrene scaffold for 3D cell culture, providing 3D space in which cells can invade, proliferate and grow
Supplier:  Stemgent™ AVP009320
 View more versions of this product
Catalog No. AVP009320
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Sterilized using gamma radiation
  • Individually sterile blister-packed plates
  • Vertical axis of 3D enables additional layers of cells to form complex 3D interactions with adjacent cells simulating structure of tissue
Specifications
Specifications
Alvetex
320
320 x 96 well plates
96