missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Reinecke Salt, ACS Reagent, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC423335000

 View more versions of this product

Catalog No. AC423335000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Reinecke salt
ACS Reagent Grade
C4H10CrN7S4·H2O
354.46
15, 8248
93.0% min.
0.05% max. (in dilute HCl)
Passes Test
500g
13573-16-5
Crystalline Powder
NH4(Cr(NH3)2(SCN)4)·H2O
MFCD00150925
Ammonium reineckate
Authentic
Glass bottle
Red

Chemical Identifiers

13573-16-5
NH4(Cr(NH3)2(SCN)4)·H2O
MFCD00150925
Reinecke salt
C4H10CrN7S4·H2O
354.46
Ammonium reineckate
ACS Reagent Grade

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Contact with acids liberates very toxic gas.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning