Please login to your online account to display your discounted pricing
Reichert™ Industrial Fluid Testing Refractometers
Designed specifically for checking ratio or concentration of water-soluble industrial fluids
$253.73 - $735.58
Specifications
Calibration Distilled Water
Accuracy ±0.2
For Use With (Application) Ideal for applications including Metalworking coolants and lubricants, heat treating fluids, hydraulic fluids, firefighting Foam and plating baths
Includes Durable Plastic carrying case, sample application pipets, calibration adjustment wrench
Temperature Compensation 15 to 38°C
View More Specs
Includes: 
  • Durable plastic carrying case
  • Sample application pipets
  • Calibration adjustment wrench
Products
Catalog Number Mfr. No. Increments Refractive Index Scale Range Price Quantity    
RE-10440
reichert™
13754000
0.5% 1.33 to 1.38 0 to 30% Each for $253.73
13-975-112
reichert™
13851000
0.2% 1.33 to 1.35 0 to 10% Each for $735.58
Description & Specifications
Specifications
Calibration Distilled Water
Accuracy ±0.2
For Use With (Application) Ideal for applications including Metalworking coolants and lubricants, heat treating fluids, hydraulic fluids, firefighting Foam and plating baths
Includes Durable Plastic carrying case, sample application pipets, calibration adjustment wrench
Temperature Compensation 15 to 38°C
Portable testers are ideal for applications including metalworking coolants and lubricants, heat-treating fluids, hydraulic fluids, firefighting foam and plating baths.
  • Automatic temperature compensation provides accurate readings without correction tables or temperature measurement
  • Temperature-compensation range, 15° to 38°C (59° to 100°F)
  • Large, easy-to-read scales and depth of field
  • Portable: no batteries or power cord required
  • Rugged, chemical-resistant housing
IFT10HP Model
  • High Precision optical instrument with improved 0-10 scale 0.1 scale increments for testing low concentration synthetic cutting and grinding fluids and lubricants
  • 10X focusable eyepiece
  • Coated antireflection glass optical components improve illumination and shadowline contrast
IFT40 Rhino Model
  • Sealed housing and eyepiece; IP67 rated water resistant and dustproof
  • Black polycarbonate housing reduces stray light
  • 0-30 arbitrary scale is divided into 0.5 increments; accuracy to ±0.25%
  • Eyepiece with large depth of focus
  • Built for production floor use; impact resistant to withstand repeated drops from three feet
One year