missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Reichert™ Reichert™ Goldberg Ultra-Precision Handheld Refractometers

Offer reading accuracy better than most benchtop laboratory refractometers

$2,465.83 - $2,465.83

Specifications

Calibration Distilled Water
Dimensions (L x W x H) 6.2 x 1 x 1 in. (157.4 x 25.4 x 25.4mm)
For Use With (Application) Ideal for use in quality control, research and clinical laboratories
Includes Soft holster case
Refractive Index 1.33 to 1.37
View More Specs

Products 2
Catalog Number Mfr. No. Accuracy Scale Range Increments Resolution Price Quantity  
Catalog Number Mfr. No. Accuracy Scale Range Increments Resolution Price Quantity  
BL-10-419 View Documents Reichert™
13104190
±0.3ppt 0 to 160ppt 0.0002nD/2ppt 0.0001 Each for $2,465.83
 
RE-10423 View Documents Reichert™
13104230
±0.1% 0 to 30% 0.1% 0.1 Each for $2,465.83
 
Description

Description

Offer reading accuracy better than most benchtop laboratory refractometers

  • Provide rapid, reliable measurements
  • For measuring Brix and salinity
  • Incorporate the Goldberg method, recognized as the most accurate and precise for handheld refractometers
  • Ideal for use in quality control, research, and clinical laboratories
  • Feature a hermetically sealed liquid-filled optical prism that changes index with temperature to compensate instantly and automatically for temperature changes
  • Heavy-duty anodized aluminum body
  • Magnified and focusable scales feature easy-to-read markings and divisions

Specifications

Specifications

Distilled Water
Ideal for use in quality control, research and clinical laboratories
1.33 to 1.37
6.2 x 1 x 1 in. (157.4 x 25.4 x 25.4mm)
Soft holster case