Reichert™ Goldberg Clinical Handheld TS Meter Refractometer
Offer reading accuracy better than most benchtop laboratory refractometers
Manufacturer:  Reichert™ 1310400A
Catalog No. 13-946-100
Description
Description
Offer reading accuracy better than most benchtop laboratory refractometers
  • Provides rapid, reliable measurements
  • For measuring urione specific gravity, serum, plasma, protein, refractive index
  • Incorporates the Goldberg method, recognized as the most accurate and precise for handheld refractometer
  • Ideal for use in quality control, research, and clinical laboratories
  • Features a hermetically sealed liquid-filled optical prism that changes index with temperature to compensate instantly and automatically for temperature changes
  • Heavy-duty anodized aluminum body
  • Magnified and focusable scales feature easy-to-read markings and divisions
Specifications
Specifications
0 to 320
±0.001 USG, ±0.1g/mL Serum Protein, ±2nD
6.2 in.
0.0001 sp. gr., 0.1g/100mL, 2 (n-nD x 104)
0.0001
1 in.
1 x 1 x 6.2 in. (25.4 x 25.4 x 157.4mm)
Clinical Handheld TS Meter Refractometer
1 to 1.035 USG, 2.5 to 15g/100mL Serum Protein, 0 to 320nD
157.4mm
1 in.
25.4mm
25.4mm