Reichert™ Auxiliary Magnifying Lens
For strain-free visual identification of colonies
Supplier:  Reichert™ 13333100
Catalog No. AO-3331
Description
Description
Specifications
Specifications
Lens, Auxiliary Magnifying
For strain-free visual identification of colonies, doubles the total magnification from 1.5X to 3.0X
Colony counter lens
1.5x