missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Regorafenib 98.0+%, TCI America™

Manufacturer:  Tci America R014225MG

 View more versions of this product

Catalog No. R014225MG



Specifications

Specifications

Regorafenib
208°C
755037-03-7
MFCD16038047
CNC(=O)C1=NC=CC(=C1)OC2=CC(=C(C=C2)NC(=O)NC3=CC(=C(C=C3)Cl)C(F)(F)F)F
482.82
CHEBI:68647
Crystal-Powder at 20°C
Undesignated
25mg
C21H15ClF4N4O3
FNHKPVJBJVTLMP-UHFFFAOYSA-N
4-[4-[[4-chloro-3-(trifluoromethyl)phenyl]carbamoylamino]-3-fluorophenoxy]-N-methylpyridine-2-carboxamide
11167602
482.82
≥98.0% (HPLC)

Chemical Identifiers

755037-03-7
482.82
FNHKPVJBJVTLMP-UHFFFAOYSA-N
CHEBI:68647
CNC(=O)C1=NC=CC(=C1)OC2=CC(=C(C=C2)NC(=O)NC3=CC(=C(C=C3)Cl)C(F)(F)F)F
C21H15ClF4N4O3
MFCD16038047
11167602
4-[4-[[4-chloro-3-(trifluoromethyl)phenyl]carbamoylamino]-3-fluorophenoxy]-N-methylpyridine-2-carboxamide
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

Documents