Please login to your online account to display your discounted pricing

Science First™ Refracting Telescope Kit


Students make a working refracting telescope

Manufacturer: science first™  PS-04B/S

 View more versions of this product

Catalog No. S48822

  • / Each
 

Description & Specifications

Specifications

Diameter (Metric) Lens 43mm and 17.5mm
Product Type Refracting telescope kit
Length (Metric) Focal 400mm and 25mm
Class Size Single kit
Includes Inner and outer tubes, plastic eyepiece lenses, foam holders, cardboard spacers and washers for the eyepiece lenses, red plastic caps and 1 set of instructions and activitie
For Use With (Application) Study how lenses work when put together without using an optical bench