Please login to your online account to display your discounted pricing

Brady™ Reflective Quik-Lite™ Ten Packs - Printed Number: 9

Identify equipment, bins and areas of warehouse required by facility safety and maintenance. Brady™ Reflective Quik-Lite™ Ten Packs can be used to mark tool cribs, bins, shelves, poles, panels, switches and equipment.

$7.98 - $19.63

Specifications

Product Type Reflective Quik-Lite, Printed Number
Display Text 9
Thickness (English) 0.013 in.
Quantity 10/Pk.
Material Reflective Vinyl Sheeting (B-957)
View More Specs
Products
Catalog Number Mfr. No. Height (English) Character Height (Metric) Character Price Quantity    

19-108-700

 
brady™
97167
1 in. Pack of 10 Each for $7.98

19-108-860

 
brady™
97445
1.5 in. Pack of 10 Each for $19.63
Description & Specifications

Specifications

Product Type Reflective Quik-Lite, Printed Number
Display Text 9
Thickness (English) 0.013 in.
Quantity 10/Pk.
Material Reflective Vinyl Sheeting (B-957)
Adhesive Type Permanent Acrylic
Color White
For Use With (Application) Labeling in Low Light Conditions; Safety and Compliance ID
Description Reflective: Engineer-Grade

  • Made of B-957 reflective material (Vinyl)
  • With permanent, aggressive adhesive grips firmly to most clean, dry surfaces, indoors and outdoors