missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Redox Standard Solution, Iodide/Triiodide, Ricca Chemical

$147.58 - $227.15

Chemical Identifiers

CAS 7732-18-5,7681-11-0,10043-35-3,7553-56-2,1310-58-3
CAS Min % 59.0,38.31,0.73,0.1,0.07
CAS Max % 61.0,39.87,0.76,0.11,0.08
Physical Form Liquid
Chemical Name or Material Redox Standard Solution, Iodide/Triiodide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
6595-16 SDS Ricca Chemical Company
RI659516
Poly natural 500mL Each for $147.58
6595-32 Ricca Chemical Company
RI659532
Poly natural 1L Each for $227.15
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7681-11-0,10043-35-3,7553-56-2,1310-58-3
61.0,39.87,0.76,0.11,0.08
Redox Standard Solution, Iodide/Triiodide
59.0,38.31,0.73,0.1,0.07
Liquid

Specifications

7732-18-5,7681-11-0,10043-35-3,7553-56-2,1310-58-3
61.0,39.87,0.76,0.11,0.08
Poly natural
500mL
59.0,38.31,0.73,0.1,0.07
Liquid
Clear brown
Redox Standard Solution, Iodide/Triiodide