Please login to your online account to display your discounted pricing

Red Rubber Sleeve Stoppers for Serum and Vaccine Bottles


Manufacturer: the plasticoid company  20200*

 View more versions of this product

Catalog No. 03-215-5

  • / Pack of 100 Each
 

Description & Specifications

Specifications

Bottom Diameter 5.16mm
Diameter (English) Bottom 0.2 in.
Diameter (English) Outside 0.26 in.
Diameter (Metric) Bottom 0.51cm
For Use With (Equipment) Serum and vaccine bottles
Length (English) Sleeve 0.42 in.
Length (Metric) Sleeve 1cm
Diameter (Metric) Outside 0.67cm
Size 11.5
Stopper Number 11.5
Color Red
Material Rubber
Product Type Sleeve Stopper