Rectangular Magnets
$57.15 - $57.15
Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
S43032 Fisher Science Education
PMCM312
Each for $57.15
Specifications
Description
A pair of Alnico, block-shaped magnets is suitable for demonstrating the extremes of attraction and repulsion. They have well-defined poles.
Order Info
They measure 26 × 19 × 13mm.