Please login to your online account to display your discounted pricing

Corning™ Rectangular Ice Pan, Maxi 9L


Multi-purpose EVA pans that provide a non-toxic alternative to conventional ice pans

Manufacturer: corning™  C432095

 View more versions of this product

Catalog No. 07-210-095

  • / Case
 

Description & Specifications

Specifications

Color Red
Capacity 9L
Dimensions (L x W x H) 40.6 x 31.8 x 11.4cm