Please login to your online account to display your discounted pricing

Corning™ Rectangular Ice Pan with Lid, Midi 4L


Multi-purpose EVA pans that provide a non-toxic alternative to conventional ice pans

Manufacturer: corning™  C432113

 View more versions of this product

Catalog No. 07-210-113

  • / Case
 

Description & Specifications

Specifications

Capacity 4L
Color Pink
Dimensions (L x W x H) 35.0 x 22.5 x 11.4cm