Please login to your online account to display your discounted pricing

Rec 15/2615 dihydrochloride, Tocris

Selective α1B antagonist

$218.74 - $915.10

Chemical Identifiers

Chemical Name or Material Rec 15/2615 dihydrochloride
Assay Percent Range >98%
Molecular Formula C26H33N5O5·2HCl
Formula Weight 568.49
Synonym 1-(4-Amino-6, 7-dimethoxy-2-quinazolinyl)-4-[[2-methoxy-6-(1-methylethyl)phenoxy]acetyl]piperazine dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

328450

 
r & d systems
328450
50mg Each for $915.10

328410

 
r & d systems
328410
10mg Each for $218.74
Description & Specifications

Specifications

Formula Weight 568.49
Quantity 50mg
Chemical Name or Material Rec 15/2615 dihydrochloride
Assay Percent Range >98%
Molecular Formula C26H33N5O5·2HCl
CAS 173059-17-1
Synonym 1-(4-Amino-6, 7-dimethoxy-2-quinazolinyl)-4-[[2-methoxy-6-(1-methylethyl)phenoxy]acetyl]piperazine dihydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature