Please login to your online account to display your discounted pricing

Reagent Alcohol, Certified, 50% (v/v), LabChem™

$37.03 - $252.31

Chemical Identifiers

Chemical Name or Material Reagent Alcohol
CAS 7732-18-5
CAS Max % 66.39
CAS 64-17-5
CAS Max % 30.25
CAS 67-63-0
CAS Max % 1.68
CAS 67-56-1
CAS Max % 1.68
Purity Grade Certified Grade
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC222201

 
lab chem
LC222201
Poly Bottle 500mL Each for $37.03

LC222204

 
lab chem
LC222204
Poly Bottle 4L Each for $75.26

LC222205

 
lab chem
LC222205
Cubitainer™ 20L Each for $252.31
Description & Specifications

Specifications

Formula Weight 46.07
Purity Grade Certified Grade
Viscosity 2.9cSt
Specific Gravity 0.928g/mL ±0.008
CAS Max % 1.68
Assay 50% (v/v)
Decomposition Information Carbon monoxide; Carbon dioxide; May release flammable gases
Solubility Soluble in water
Chemical Name or Material Reagent Alcohol
CAS 64-17-5
EINECS Number 200-578-6
Packaging Poly Bottle
Synonym Ethyl Alcohol; SDA 3A
Color Clear, colorless
CAS 7732-18-5
CAS Max % 30.25
Identification Passes Test
Density 0.95g/mL
CAS 67-56-1
Health Hazard 1 Danger
Physical Form Liquid
CAS 67-63-0
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, hot surfaces, open flames, sparks.
No smoking.
Keep container tightly closed.
Keep cool.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, lighting, ventilating equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, spray, vapors.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If skin irritation occurs: Get medical advice/attention.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
In case of fire: Use carbon dioxide (CO2), powder, alcohol-resistant foam to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
UN Number UN1987
CAS Max % 1.68
CAS Max % 66.39
Storage Information Room Temperature
Health Hazard 2 GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Suspected of damaging fertility or the unborn child.
Causes damage to organs (central nervous system, optic nerve) (oral, dermal).
Quantity 500mL
Linear Formula C2H6O
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, hot surfaces, open flames, sparks.
No smoking.
Keep container tightly closed.
Keep cool.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, lighting, ventilating equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, spray, vapors.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If skin irritation occurs: Get medical advice/attention.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
In case of fire: Use carbon dioxide (CO2), powder, alcohol-resistant foam to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Suspected of damaging fertility or the unborn child.
Causes damage to organs (central nervous system, optic nerve) (oral, dermal).